Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP7D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173516.1 | NP_001166987.1 | 4 | 438 | ||
NM_001173517.1 | NP_001166988.1 | 4 | 550 | |||
NM_024597.3 | NP_078873.2 | 4 | 550 | |||
XM_005262472.1 | XP_005262529.1 | 4 | 405 | |||
XM_017029843.1 | XP_016885332.1 | 4 | 402 | |||
GenBank mRNA | AK301478.1 | - | 4 | 439 | ||
AL832120.1 | - | 4 | 342 | |||
BC040518.1 | - | 4 | 414 | |||
BC064350.1 | - | 4 | 353 |