Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098802.1 | NP_001092272.1 | 5 | 975 | ||
NM_032171.1 | NP_115547.1 | 5 | 975 | |||
XM_005252263.4 | XP_005252320.1 | 5 | 988 | |||
XM_005252265.2 | XP_005252322.1 | 5 | 988 | |||
XM_005252266.4 | XP_005252323.1 | 5 | 982 | |||
XM_017015192.1 | XP_016870681.1 | 5 | 988 | |||
XM_017015193.1 | XP_016870682.1 | 5 | 988 | |||
XM_017015194.1 | XP_016870683.1 | 5 | 988 | |||
XM_017015195.1 | XP_016870684.1 | 5 | 988 | |||
XM_017015196.1 | XP_016870685.1 | 5 | 982 | |||
GenBank mRNA | AK303457.1 | - | 5 | 1025 | ||
BC058931.1 | - | 5 | 888 | |||
BC128058.1 | - | 5 | 843 | 40078333 |