Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206998.1 | NP_001193927.1 | 5 | 645 | ||
NM_032173.3 | NP_115549.2 | 5 | 438 | |||
XM_011530435.2 | XP_011528737.1 | 6 | 634 | |||
XM_011530436.2 | XP_011528738.1 | 7 | 689 | |||
XM_011530437.1 | XP_011528739.1 | 6 | 1359 | |||
XM_011530438.2 | XP_011528740.1 | 6 | 875 | |||
XM_017028990.1 | XP_016884479.1 | 6 | 1130 | |||
GenBank mRNA | AB051436.1 | - | 5 | 510 | ||
AK096397.1 | - | 5 | 438 | |||
CR456397.1 | - | 5 | 418 |