Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000061.1 | - | 9 | 1414 | ||
AK024832.1 | - | 2 | 761 | |||
AK091207.1 | - | 20 | 2438 |