Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080495.2 | NP_001073964.2 | 6, 7 | 2604 | ||
XM_017012728.1 | XP_016868217.1 | 6, 7 | 5101 | |||
XM_017012729.1 | XP_016868218.1 | 5, 6 | 2808 | |||
XM_017012730.1 | XP_016868219.1 | 5, 6 | 2426 | |||
XM_017012731.1 | XP_016868220.1 | 5, 6 | 2427 | |||
XM_017012732.1 | XP_016868221.1 | 5, 6 | 2427 | |||
XM_017012733.1 | XP_016868222.1 | 4, 5 | 2283 | |||
XM_017012734.1 | XP_016868223.1 | 5, 6 | 3309 | |||
XM_017012735.1 | XP_016868224.1 | 5, 6 | 2311 | |||
XM_017012736.1 | XP_016868225.1 | 5, 6 | 2898 | |||
XM_017012737.1 | XP_016868226.1 | 5, 6 | 4671 | |||
XM_017012738.1 | XP_016868227.1 | 5, 6 | 2193 | |||
XM_017012739.1 | XP_016868228.1 | 5, 6 | 2428 | |||
XM_017012740.1 | XP_016868229.1 | 5, 6 | 2428 | |||
GenBank mRNA | CN295035.1 | - | 2, 3 | 203 | ||
CR993399.1 | - | 2, 3 | 461 |