Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304360.1 | NP_001291289.1 | 11 | 1348 | ||
XM_011542333.2 | XP_011540635.2 | 11 | 1265 | |||
XM_011542335.1 | XP_011540637.1 | 6 | 777 | |||
XM_011542336.2 | XP_011540638.1 | 6 | 2512 | |||
XM_017002641.1 | XP_016858130.1 | 12 | 1474 | |||
XM_017002642.1 | XP_016858131.1 | 12 | 1474 | |||
XM_017002643.1 | XP_016858132.1 | 12 | 1474 | |||
GenBank mRNA | AB051538.1 | - | 8 | 1020 | ||
BC139754.1 | - | 10 | 1225 |