Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIPPLY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171706.1 | NP_001165177.1 | 2 | 857 | ||
NM_138382.2 | NP_612391.1 | 4 | 998 | |||
GenBank mRNA | BC007652.1 | - | 4 | 984 | ||
BC101998.1 | - | 4 | 998 | |||
BC105691.1 | - | 4 | 998 | 40023299 | ||
BC105692.1 | - | 4 | 998 | 40023298 | ||
BC110436.1 | - | 3 | 1915 | 40023301 | ||
BC127250.1 | - | 2 | 857 | 40023300 |