Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080426.1 | NP_001073895.1 | 5 | 3407 | ||
XM_005245603.3 | XP_005245660.1 | 6 | 3570 | |||
XM_011510146.2 | XP_011508448.1 | 5 | 3444 | |||
GenBank mRNA | AF119045.1 | - | 1 | 1916 | ||
AK094814.1 | - | 1 | 2004 | |||
BC115387.1 | - | 6 | 3527 |