Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002607.5 | NP_002598.4 | 7 | 2261 | ||
NM_033023.4 | NP_148983.1 | 6 | 2192 | |||
XM_011515415.1 | XP_011513717.1 | 8 | 1753 | |||
XM_011515416.1 | XP_011513718.1 | 8 | 1757 | |||
XM_011515417.1 | XP_011513719.1 | 7 | 1684 | |||
XM_011515418.1 | XP_011513720.1 | 7 | 1688 | |||
XM_011515419.2 | XP_011513721.1 | 7 | 1461 | |||
XM_017012289.1 | XP_016867778.1 | 7 | 1494 | |||
GenBank mRNA | X06374.1 | - | 6 | 1762 |