Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303012.1 | NP_001289941.1 | 11 | 1611 | ||
NM_001303013.1 | NP_001289942.1 | 11 | 2797 | |||
NM_014638.3 | NP_055453.2 | 11 | 1805 | |||
XM_011542455.2 | XP_011540757.1 | 12 | 1847 | |||
XM_011542456.2 | XP_011540758.1 | 12 | 1804 | |||
XM_017002870.1 | XP_016858359.1 | 12 | 1836 | |||
XM_017002871.1 | XP_016858360.1 | 12 | 1807 | |||
XM_017002872.1 | XP_016858361.1 | 12 | 1836 | |||
XM_017002873.1 | XP_016858362.1 | 12 | 1836 | |||
XM_017002874.1 | XP_016858363.1 | 12 | 1836 | |||
GenBank mRNA | AB007919.2 | - | 11 | 1611 | ||
AK122589.1 | - | 9 | 1904 | |||
BC043358.1 | - | 9 | 1107 | |||
BC128207.1 | - | 9 | 1187 | |||
BC144620.1 | - | 11 | 2789 | |||
DQ176850.1 | - | 11 | 1805 |