Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184998.1 | NP_001171927.1 | 27 | 7462 | ||
NM_001184999.1 | NP_001171928.1 | 27 | 7453 | |||
NM_014647.3 | NP_055462.2 | 27 | 7462 | |||
XM_005255704.1 | XP_005255761.1 | 27 | 7458 | |||
XM_005255706.1 | XP_005255763.1 | 27 | 7452 | |||
XM_005255707.1 | XP_005255764.1 | 27 | 7428 | |||
XM_017023900.1 | XP_016879389.1 | 27 | 7346 | |||
XM_017023901.1 | XP_016879390.1 | 27 | 7343 | |||
XM_017023902.1 | XP_016879391.1 | 26 | 7296 | |||
XM_017023903.1 | XP_016879392.1 | 26 | 7029 | |||
XM_017023904.1 | XP_016879393.1 | 26 | 7026 | |||
XM_017023905.1 | XP_016879394.1 | 27 | 6883 | |||
XM_017023906.1 | XP_016879395.1 | 27 | 6824 | |||
GenBank mRNA | AB007890.3 | - | 25 | 6548 | ||
AK124564.1 | - | 3 | 3878 |