Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLURP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020427.2 | NP_065160.1 | 3 | 306 | ||
GenBank mRNA | AY579079.1 | - | 3 | 306 | ||
AY579080.1 | - | 3 | 306 | |||
BC069292.1 | - | 3 | 306 | 7262730 | ||
BC105133.1 | - | 3 | 303 | 8144136 | ||
BC105135.1 | - | 3 | 301 | 8144138 |