Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039675.1 | NP_001034764.1 | 8, 9 | 1311 | ||
NM_001323619.1 | NP_001310548.1 | 5, 6 | 570 | |||
NM_001323620.1 | NP_001310549.1 | 5, 6 | 1108 | |||
NM_001323621.1 | NP_001310550.1 | 5, 6 | 915 | |||
NM_018671.4 | NP_061141.2 | 5, 6 | 960 | |||
XM_005254963.3 | XP_005255020.1 | 5, 6 | 680 | |||
XM_011521779.2 | XP_011520081.1 | 8, 9 | 1556 | |||
XM_011521780.2 | XP_011520082.1 | 4, 5 | 462 | |||
XM_017022407.1 | XP_016877896.1 | 5, 6 | 680 | |||
GenBank mRNA | AB014729.1 | - | 5, 6 | 502 | ||
AB014736.1 | - | 5, 6 | 544 | |||
AK000905.1 | - | 4, 5 | 838 | |||
AK125721.1 | - | 8, 9 | 1311 | |||
AK291622.1 | - | 5, 6 | 515 | |||
AK293538.1 | - | 5, 6 | 684 | |||
AK296964.1 | - | 5, 6 | 537 | |||
AK302979.1 | - | 5, 6 | 577 | |||
BC006214.1 | - | 5, 6 | 543 | 3051384 | ||
BC037992.1 | - | 5, 6 | 543 | 5534906 | ||
BC045635.1 | - | 8, 9 | 1311 | 5297091 |