Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033583.2 | NP_001028755.2 | 8, 9 | 788 | ||
NM_001037171.1 | NP_001032248.1 | 9, 10 | 815 | |||
XM_005274472.2 | XP_005274529.1 | 7, 8 | 687 | |||
GenBank mRNA | AF132950.1 | - | 7, 8 | 690 | ||
AK000659.1 | - | 8, 9 | 774 | |||
AK024337.1 | - | 10, 11 | 1880 | |||
AL548456.3 | - | 9, 10 | 815 | |||
BC136671.1 | - | 8, 9 | 739 | 9020661 | ||
BC144360.1 | - | 8, 9 | 739 | 9052881 | ||
BX648512.1 | - | 9, 10 | 799 |