Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033583.2 | NP_001028755.2 | 2 | 250 | ||
NM_001037171.1 | NP_001032248.1 | 2 | 250 | |||
GenBank mRNA | AK000659.1 | - | 2 | 236 | ||
AK024337.1 | - | 4 | 1342 | |||
AL548456.3 | - | 2 | 250 | |||
BC012573.1 | - | 2 | 221 | |||
BC136671.1 | - | 2 | 201 | 9020661 | ||
BC144360.1 | - | 2 | 201 | 9052881 | ||
BC144370.1 | - | 2 | 201 | |||
BX648512.1 | - | 2 | 234 |