Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C17ORF62 SIRNA Assays ›
Gene Symbol:
C17orf62
Gene Name:
chromosome 17 open reading frame 62
Gene Aliases:
-
Chromosome Location:
Chr.17: 82442586 - 82450831 on Build GRCh38
UniGene ID:
Hs.163113
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
279201
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C17orf62
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033046.3
NP_001028218.1
5
399
NM_001100407.2
NP_001093877.1
6
449
NM_001100408.2
NP_001093878.1
4
357
NM_001193653.1
NP_001180582.1
6
669
NM_001193654.1
NP_001180583.1
5
619
NM_001193655.1
NP_001180584.1
5
563
NM_001193657.1
NP_001180586.1
6
616
NR_036514.1
4
325
NR_036516.1
4
276
NR_036517.1
3
234
NR_036518.1
2
160
XM_006722293.2
XP_006722356.1
4
1124
XM_011523606.2
XP_011521908.1
4
1304
XM_017025072.1
XP_016880561.1
4
1304
XM_017025073.1
XP_016880562.1
3
1262
XM_017025074.1
XP_016880563.1
5
397
XM_017025075.1
XP_016880564.1
5
1166
XM_017025076.1
XP_016880565.1
6
394
XM_017025077.1
XP_016880566.1
5
558
XM_017025078.1
XP_016880567.1
4
355
GenBank mRNA
AK027743.1
-
2
100
AK054876.1
-
4
355
AK056342.1
-
4
232
AK074950.1
-
6
546
AK090484.1
-
4
2470
AK092865.1
-
2
1458
AK309201.1
-
5
397
AU280357.1
-
5
344
BC003595.1
-
5
283
2822930
BC004171.2
-
5
283
2822930
BG338078.1
-
5
564
BG716477.1
-
5
499
BM147030.1
-
4
181
BM801484.1
-
6
382
CN260096.1
-
4
183
DA008508.1
-
3
131
DA342707.1
-
4
347
DA500882.1
-
4
281
DA663677.1
-
6
371
DA716299.1
-
5
403
DA969007.1
-
6
493
DB113745.1
-
4
180
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0