Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033046.3 | NP_001028218.1 | 7 | 1666 | ||
NM_001100407.2 | NP_001093877.1 | 8 | 1716 | |||
NM_001100408.2 | NP_001093878.1 | 6 | 1624 | |||
NM_001193653.1 | NP_001180582.1 | 8 | 1936 | |||
NM_001193654.1 | NP_001180583.1 | 7 | 1886 | |||
NM_001193655.1 | NP_001180584.1 | 7 | 1830 | |||
NM_001193657.1 | NP_001180586.1 | 8 | 1883 | |||
NR_036514.1 | 6 | 1592 | ||||
NR_036516.1 | 6 | 1543 | ||||
NR_036517.1 | 5 | 1501 | ||||
NR_036518.1 | 4 | 1427 | ||||
XM_006722293.2 | XP_006722356.1 | 6 | 2391 | |||
XM_011523606.2 | XP_011521908.1 | 7 | 2523 | |||
XM_017025072.1 | XP_016880561.1 | 6 | 2571 | |||
XM_017025073.1 | XP_016880562.1 | 5 | 2529 | |||
XM_017025074.1 | XP_016880563.1 | 8 | 1616 | |||
XM_017025075.1 | XP_016880564.1 | 8 | 2385 | |||
XM_017025076.1 | XP_016880565.1 | 9 | 1613 | |||
XM_017025077.1 | XP_016880566.1 | 8 | 1777 | |||
XM_017025078.1 | XP_016880567.1 | 7 | 1574 | |||
GenBank mRNA | AF269290.1 | - | 2 | 1789 | ||
AK054876.1 | - | 7 | 1478 | |||
AK074950.1 | - | 8 | 1813 | |||
AK090484.1 | - | 6 | 3737 | |||
AK092865.1 | - | 4 | 2725 | |||
AK225389.1 | - | 7 | 1572 | |||
BC003595.1 | - | 7 | 1550 | 2822930 | ||
BC004171.2 | - | 7 | 1550 | 2822930 |