Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018124.3 | NP_060594.3 | 13 | 4398 | ||
XM_005256021.4 | XP_005256078.1 | 14 | 4531 | |||
XM_005256022.4 | XP_005256079.1 | 13 | 5281 | |||
XM_006721228.3 | XP_006721291.1 | 13 | 4354 | |||
XM_011523191.2 | XP_011521493.1 | 13 | 4351 | |||
XM_017023391.1 | XP_016878880.1 | 14 | 4447 | |||
XM_017023392.1 | XP_016878881.1 | 12 | 4218 | |||
GenBank mRNA | AK022673.1 | - | 3 | 2437 | ||
BC002574.2 | - | 11 | 3775 |