Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014817.3 | NP_055632.2 | 1 | 4916 | ||
GenBank mRNA | AB014544.1 | - | 1 | 4795 |