Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023570.3 | NP_001018864.2 | 11 | 1324 | ||
NM_001023571.3 | NP_001018865.2 | 8 | 925 | |||
NM_001319107.1 | NP_001306036.1 | 11 | 1323 | |||
NR_134968.1 | 10 | 1213 | ||||
XM_005247911.3 | XP_005247968.1 | 11 | 1323 | |||
XM_005247912.2 | XP_005247969.1 | 10 | 1137 | |||
XM_011513335.2 | XP_011511637.1 | 9 | 1043 | |||
XM_017007537.1 | XP_016863026.1 | 10 | 1206 | |||
XM_017007538.1 | XP_016863027.1 | 7 | 1008 | |||
XM_017007539.1 | XP_016863028.1 | 8 | 911 | |||
GenBank mRNA | AB062481.1 | - | 10 | 1207 | ||
AY714228.1 | - | 11 | 1318 | |||
AY964667.1 | - | 8 | 875 | |||
AY964668.1 | - | 11 | 1262 | |||
BC005806.2 | - | 8 | 866 | 3834623 | ||
D25278.1 | - | 11 | 1265 |