Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MZT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025029.3 | NP_079305.2 | 3 | 875 | ||
XM_005263795.4 | XP_005263852.1 | 2 | 780 | |||
XM_011511886.2 | XP_011510188.1 | 4 | 799 | |||
XM_011511887.2 | XP_011510189.1 | 4 | 766 | |||
XM_011511888.2 | XP_011510190.1 | 4 | 1100 | |||
GenBank mRNA | BC014255.2 | - | 3 | 517 | ||
BC066296.1 | - | 3 | 607 | 5284352 |
Gene Symbol: MZT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085365.1 | NP_001078834.1 | 3 | 635 | ||
GenBank mRNA | BC018206.1 | - | 3 | 539 | 3862861 | |
BC063024.1 | - | 3 | 518 | |||
BC104650.1 | - | 2 | 416 | |||
BM554057.1 | - | 3 | 697 |
Gene Symbol: PHGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FJ224347.1 | - | 543 |