Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MZT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025029.3 | NP_079305.2 | 3 | 913 | ||
XM_005263795.4 | XP_005263852.1 | 2 | 818 | |||
XM_011511886.2 | XP_011510188.1 | 4 | 837 | |||
XM_011511887.2 | XP_011510189.1 | 4 | 804 | |||
XM_011511888.2 | XP_011510190.1 | 4 | 1138 | |||
GenBank mRNA | BC014255.2 | - | 3 | 555 |
Gene Symbol: MZT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085365.1 | NP_001078834.1 | 3 | 673 | ||
GenBank mRNA | BM554057.1 | - | 3 | 735 |