Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004740.3 | NP_004731.2 | 1, 2 | 2082 | ||
GenBank mRNA | AF105277.1 | - | 791 | |||
BC047669.1 | - | 1, 2 | 1379 | 6056136 | ||
BC058075.1 | - | 1065 |