Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020991.3 | NP_066271.1 | 5 | 757 | ||
NM_022644.3 | NP_072170.1 | 4 | 1010 | |||
NM_022645.2 | NP_072171.1 | 3 | 472 | |||
GenBank mRNA | BC022044.1 | - | 5 | 708 | 4769046 | |
BC035965.1 | - | 5 | 704 | 4731880 | ||
BC119748.2 | - | 5 | 701 | 40119242 | ||
V00573.1 | - | 5 | 666 |