Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005675.4 | NP_005666.2 | 5 | 1200 | ||
XM_011530394.1 | XP_011528696.1 | 6 | 1187 | |||
GenBank mRNA | AK098780.1 | - | 5 | 1422 | ||
BC047039.1 | - | 5 | 1093 | 5229172 |
Gene Symbol: DGCR6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033257.3 | NP_150282.2 | 5 | 1165 | ||
GenBank mRNA | AK001528.1 | - | 5 | 1106 | ||
AK289952.1 | - | 5 | 1083 | |||
AK307210.1 | - | 4 | 1000 | |||
BC000682.2 | - | 5 | 1101 | 3349309 |
Gene Symbol: LOC102724770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724934.1 | XP_006724997.1 | 5 | 1142 |