Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027250.2 | 3 | 369 | |||
GenBank mRNA | AK302291.1 | - | 3 | 370 | ||
DC399111.1 | - | 3 | 370 |