Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032391.1 | NP_001027563.1 | 9 | 1016 | ||
NM_016309.2 | NP_057393.2 | 11 | 1181 | |||
XM_005255354.3 | XP_005255411.1 | 11 | 970 | |||
XM_011545862.2 | XP_011544164.1 | 10 | 1130 | |||
XM_011545864.1 | XP_011544166.1 | 12 | 1295 | |||
GenBank mRNA | AF151826.1 | - | 11 | 1165 | ||
AK025884.1 | - | 12 | 1288 | |||
AK223364.1 | - | 12 | 1339 | |||
AK225271.1 | - | 11 | 1125 | |||
AK291885.1 | - | 11 | 1125 | |||
AL137283.1 | - | 11 | 985 | |||
BC001214.1 | - | 11 | 1148 | 3533656 | ||
BC014217.2 | - | 12 | 1269 | 3959936 |