Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019102.3 | NP_061975.2 | 2 | 943 | ||
GenBank mRNA | BC013682.1 | - | 2 | 930 | 3863578 |