Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005259057.2 | XP_005259114.1 | 11, 12 | 1560 | ||
XM_011527113.1 | XP_011525415.1 | 11, 12 | 1315 | |||
XM_011527114.1 | XP_011525416.1 | 11, 12 | 1253 | |||
XM_011527115.1 | XP_011525417.1 | 11, 12 | 1321 | |||
XM_011527116.1 | XP_011525418.1 | 11, 12 | 1447 | |||
XM_017026988.1 | XP_016882477.1 | 7, 8 | 671 | |||
GenBank mRNA | AK022948.1 | - | 11, 12 | 1532 | ||
AL136606.1 | - | 11, 12 | 1486 | |||
BC008869.2 | - | 11, 12 | 1472 | 4131601 |