Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FBLIM1 SIRNA Assays ›
Gene Symbol:
FBLIM1
Gene Name:
filamin binding LIM protein 1
Gene Aliases:
CAL, FBLP-1, FBLP1
Chromosome Location:
Chr.1: 15756171 - 15786590 on Build GRCh38
UniGene ID:
Hs.530101
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
279294
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FBLIM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001024215.1
NP_001019386.1
5
880
NM_001024216.1
NP_001019387.1
2
402
NM_017556.2
NP_060026.2
6
972
XM_005245900.1
XP_005245957.1
6
974
XM_005245901.1
XP_005245958.1
6
940
XM_005245902.1
XP_005245959.1
7
1162
XM_005245903.1
XP_005245960.1
7
1183
XM_005245909.3
XP_005245966.1
7
1284
XM_006710704.3
XP_006710767.1
7
1602
XM_006710705.1
XP_006710768.1
7
1194
XM_011541616.2
XP_011539918.1
7
1599
XM_011541617.2
XP_011539919.1
7
1183
XM_011541618.2
XP_011539920.1
7
1183
XM_017001519.1
XP_016857008.1
7
1489
XM_017001520.1
XP_016857009.1
7
1466
XM_017001521.1
XP_016857010.1
7
1492
XM_017001522.1
XP_016857011.1
7
1463
XM_017001523.1
XP_016857012.1
6
839
XM_017001524.1
XP_016857013.1
6
810
XM_017001525.1
XP_016857014.1
7
1206
XM_017001526.1
XP_016857015.1
7
1183
GenBank mRNA
AF459643.1
-
5
880
AK027444.1
-
2
420
AK055259.1
-
6
1313
AL133035.1
-
2
155
AY180161.1
-
4
673
BC019895.1
-
7
1116
5139869
CN278745.1
-
1
53
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase modulator
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell shape
single organismal cell-cell adhesion
regulation of integrin activation
cell junction assembly
protein binding
zinc ion binding
filamin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-zpscg:80/10.239.160.225:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0