Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM6SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001524.2 | NP_001001524.2 | 8, 9 | 856 | ||
GenBank mRNA | AF255923.1 | - | 8, 9 | 805 | ||
AK026307.1 | - | 5, 6 | 1140 | |||
AK097070.1 | - | 5, 6 | 1202 | |||
BC120986.1 | - | 8, 9 | 830 | |||
BC120987.1 | - | 8, 9 | 830 |