Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLYWCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142499.1 | NP_001135971.1 | 3 | 354 | ||
NM_001142500.1 | NP_001135972.1 | 3 | 444 | |||
NM_138439.2 | NP_612448.1 | 3 | 448 | |||
XM_005255078.4 | XP_005255135.1 | 2 | 651 | |||
GenBank mRNA | AL542248.3 | - | 3 | 336 | ||
BC014089.2 | - | 3 | 326 | 4330613 | ||
BG120460.1 | - | 3 | 445 | |||
BI825861.1 | - | 3 | 408 | |||
DB455461.1 | - | 3 | 428 |