Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLYWCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142499.1 | NP_001135971.1 | 4 | 709 | ||
NM_001142500.1 | NP_001135972.1 | 4 | 799 | |||
NM_138439.2 | NP_612448.1 | 4 | 803 | |||
XM_005255078.4 | XP_005255135.1 | 3 | 1006 | |||
GenBank mRNA | BC014089.2 | - | 4 | 681 | 4330613 | |
BI825861.1 | - | 4 | 761 |