Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000213.4 | NP_000204.3 | 10 | 1455 | ||
NM_001005619.1 | NP_001005619.1 | 9 | 1168 | |||
NM_001005731.2 | NP_001005731.1 | 10 | 1455 | |||
NM_001321123.1 | NP_001308052.1 | 10 | 1406 | |||
XM_005257309.2 | XP_005257366.1 | 10 | 1225 | |||
XM_005257311.4 | XP_005257368.1 | 10 | 1444 | |||
XM_006721866.3 | XP_006721929.1 | 10 | 1371 | |||
XM_006721867.3 | XP_006721930.1 | 10 | 1368 | |||
XM_006721868.3 | XP_006721931.1 | 10 | 1365 | |||
XM_006721870.3 | XP_006721933.1 | 10 | 1361 | |||
XM_011524751.2 | XP_011523053.1 | 10 | 1369 | |||
GenBank mRNA | AB208913.1 | - | 10 | 1257 | ||
AF011375.1 | - | 9 | 1160 | |||
AK310952.1 | - | 10 | 1225 | |||
BC109238.1 | - | 6 | 913 | |||
BC118916.1 | - | 10 | 1217 | 40073279 | ||
BC126411.1 | - | 10 | 1276 | 8992127 | ||
BC143738.1 | - | 10 | 1276 | 9052252 | ||
BC143742.1 | - | 10 | 1227 | 9052256 | ||
X51841.1 | - | 10 | 1286 | |||
X52186.1 | - | 9 | 1169 | |||
X53587.1 | - | 10 | 1347 |