Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 20 | 2682 | ||
NM_183372.5 | NP_899228.4 | 22 | 3376 | |||
NR_046188.3 | 23 | 3444 | ||||
NR_110864.1 | 21 | 3368 | ||||
GenBank mRNA | AK302413.1 | - | 19 | 2518 | ||
AL832622.2 | - | 23 | 3579 | |||
BC136292.1 | - | 22 | 3376 | 9020279 | ||
BX648497.1 | - | 21 | 3167 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY894571.1 | - | 17 | 2426 |