Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317801.1 | NP_001304730.1 | 4 | 414 | ||
NM_033520.2 | NP_277055.1 | 4 | 355 | |||
GenBank mRNA | AB038317.1 | - | 4 | 352 | ||
AB038318.1 | - | 4 | 411 | |||
AF213678.1 | - | 4 | 355 | |||
BC008888.2 | - | 4 | 312 | |||
BC014264.2 | - | 4 | 312 | 4550805 | ||
BC060319.1 | - | 4 | 366 | 3901226 | ||
BX648823.1 | - | 4 | 316 | |||
CB109881.1 | - | 4 | 260 |