Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015268.3 | NP_056083.3 | 56 | 6863 | ||
XM_005247245.4 | XP_005247302.1 | 57 | 7060 | |||
XM_017006036.1 | XP_016861525.1 | 43 | 5320 | |||
GenBank mRNA | AB014578.2 | - | 56 | 6912 | ||
AK023168.1 | - | 25 | 3291 | |||
AK125330.1 | - | 33 | 4247 | |||
AK127112.1 | - | 40 | 4951 | |||
AY779857.1 | - | 56 | 6863 | |||
BC000164.3 | - | 9 | 1226 | |||
BC009630.1 | - | 11 | 1346 | |||
BC040638.2 | - | 6 | 914 | |||
BC065756.1 | - | 5 | 622 | |||
BC105580.1 | - | 4 | 439 | |||
BC151246.1 | - | 57 | 6933 | 9007477 |