Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015268.3 | NP_056083.3 | 41 | 4916 | ||
XM_005247245.4 | XP_005247302.1 | 42 | 5113 | |||
XM_017006036.1 | XP_016861525.1 | 28 | 3373 | |||
GenBank mRNA | AB014578.2 | - | 41 | 4965 | ||
AK023168.1 | - | 10 | 1344 | |||
AK125330.1 | - | 18 | 2300 | |||
AK127112.1 | - | 25 | 3004 | |||
AY779857.1 | - | 41 | 4916 | |||
BC151246.1 | - | 42 | 4986 | 9007477 |