Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP10-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB076347.1 | - | 1 | 527 | ||
BC120959.2 | - | 1 | 521 | 40115271 | ||
BC120960.1 | - | 1 | 521 | 40115274 |
Gene Symbol: KRTAP10-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198693.3 | NP_941966.1 | 1 | 526 | ||
GenBank mRNA | AB076348.1 | - | 1 | 526 |