Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 4 | 2765 | ||
NR_036599.1 | 5 | 2883 | ||||
NR_036600.1 | 3 | 2638 | ||||
XM_006723354.3 | XP_006723417.1 | 4 | 2740 | |||
XM_011527262.2 | XP_011525564.1 | 4 | 2795 | |||
GenBank mRNA | BC140720.1 | - | 3 | 2666 | 9021588 |