Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTCH2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203458.4 | NP_982283.2 | 5 | 1025 | ||
GenBank mRNA | BC010154.2 | - | 5 | 1019 | 3623163 | |
BC019835.1 | - | 5 | 996 | |||
BC065219.1 | - | 5 | 977 | 5724338 | ||
BC084572.1 | - | 5 | 979 | |||
BC090943.1 | - | 5 | 995 | 5404327 | ||
BX537434.1 | - | 6 | 1104 |
Gene Symbol: LOC100996717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003047.1 | XP_016858536.1 | 5 | 741 | ||
XM_017003048.1 | XP_016858537.1 | 5 | 7436 | |||
XM_017003049.1 | XP_016858538.1 | 5 | 753 | |||
XM_017003050.1 | XP_016858539.1 | 5 | 987 | |||
XM_017003051.1 | XP_016858540.1 | 5 | 986 | |||
XM_017003052.1 | XP_016858541.1 | 6 | 887 |
Gene Symbol: LOC100996763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006711700.3 | XP_006711763.1 | 5 | 934 | ||
XM_006711703.3 | XP_006711766.1 | 5 | 996 | |||
XM_011510256.2 | XP_011508558.1 | 5 | 1721 |
Gene Symbol: LOC101929796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003012.1 | XP_016858501.1 | 5 | 1020 |