Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100996717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003051.1 | XP_016858540.1 | 5 | 1548 |
Gene Symbol: LOC100996763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006711700.3 | XP_006711763.1 | 5 | 1496 | ||
XM_006711703.3 | XP_006711766.1 | 5 | 1558 |
Gene Symbol: NOTCH2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203458.4 | NP_982283.2 | 5 | 1587 | ||
GenBank mRNA | BC065219.1 | - | 5 | 1539 | 5724338 | |
BX537434.1 | - | 6 | 1666 |