Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032172.2 | NP_115548.1 | 18 | 4728 | ||
XM_006715790.1 | XP_006715853.1 | 17 | 4631 | |||
XM_006715791.2 | XP_006715854.1 | 20 | 5137 | |||
XM_011515573.1 | XP_011513875.1 | 21 | 5196 | |||
XM_011515574.1 | XP_011513876.1 | 18 | 4720 | |||
XM_011515577.1 | XP_011513879.1 | 16 | 4349 | |||
XM_011515578.1 | XP_011513880.1 | 18 | 4771 | |||
GenBank mRNA | AA936550.1 | - | 1 | 444 | ||
AK309375.1 | - | 15 | 2315 | |||
BU567897.1 | - | 1 | 340 |