Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010969.3 | NP_001010969.2 | 6 | 764 | ||
GenBank mRNA | AY280372.1 | - | 6 | 703 | ||
BC142607.1 | - | 6 | 733 | 40147121 | ||
BC148248.1 | - | 6 | 764 | 40132876 |