Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010969.3 | NP_001010969.2 | 4 | 522 | ||
NM_001308102.1 | NP_001295031.1 | 4 | 522 | |||
XM_005270767.3 | XP_005270824.1 | 4 | 504 | |||
XM_005270770.3 | XP_005270827.1 | 4 | 504 | |||
GenBank mRNA | BC148248.1 | - | 4 | 522 | 40132876 |