Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001712.2 | NP_001001712.2 | 2 | 277 | ||
GenBank mRNA | AK091788.1 | - | 1 | 88 | ||
AY301271.1 | - | 2 | 221 | |||
BC036875.1 | - | 1 | 97 | |||
BC040963.1 | - | 1 | 1440 | |||
BC064596.1 | - | 1 | 60 | |||
BC133045.1 | - | 2 | 277 | 40147067 | ||
BC144440.1 | - | 2 | 277 | |||
BC144441.1 | - | 2 | 277 | 9052964 | ||
BC171741.1 | - | 2 | 215 |