Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001666.3 | NP_001001666.1 | 4 | 509 | ||
NM_001001891.3 | NP_001001891.2 | 4 | 512 | |||
XM_011511263.2 | XP_011509565.1 | 4 | 460 | |||
XM_011511267.2 | XP_011509569.1 | 4 | 468 | |||
XM_017004229.1 | XP_016859718.1 | 4 | 470 | |||
XM_017004230.1 | XP_016859719.1 | 3 | 363 | |||
GenBank mRNA | AY617079.1 | - | 4 | 513 | ||
AY617080.1 | - | 4 | 510 |