Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2L8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001963.1 | NP_001001963.1 | 1 | 665 | ||
GenBank mRNA | BC140751.1 | - | 1 | 746 | 9021619 |