Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008801.1 | NP_001008801.1 | 4 | 3570 | ||
NM_001277120.1 | NP_001264049.1 | 5 | 3688 | |||
NR_102299.1 | 6 | 3943 | ||||
NR_102300.1 | 6 | 3924 | ||||
NR_102301.1 | 5 | 3825 | ||||
XM_017027443.1 | XP_016882932.1 | 5 | 3785 | |||
XM_017027444.1 | XP_016882933.1 | 6 | 3903 | |||
GenBank mRNA | AK023558.1 | - | 1 | 1626 | ||
AK025501.1 | - | 1 | 2208 | |||
AY442330.1 | - | 4 | 3567 | |||
AY442331.1 | - | 5 | 3685 | |||
BC063143.1 | - | 1 | 1871 | |||
CR749381.1 | - | 1 | 2665 | |||
CR936599.1 | - | 4 | 3535 | |||
CR936695.1 | - | 4 | 3539 |