Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308169.1 | NP_001295098.1 | 5 | 878 | ||
NM_001308172.1 | NP_001295101.1 | 6 | 1063 | |||
NM_001308954.1 | NP_001295883.1 | 7 | 1445 | |||
XM_017022923.1 | XP_016878412.1 | 7 | 1038 | |||
XM_017022924.1 | XP_016878413.1 | 6 | 948 | |||
XM_017022925.1 | XP_016878414.1 | 5 | 835 | |||
XM_017022926.1 | XP_016878415.1 | 4 | 580 | |||
GenBank mRNA | AK055867.1 | - | 7 | 859 | ||
AK096039.1 | - | 7 | 1304 | |||
AK298359.1 | - | 5 | 767 | |||
AK298434.1 | - | 7 | 1060 | |||
AK303774.1 | - | 5 | 763 | |||
BC125176.1 | - | 6 | 900 | 40121949 |